To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:IP17|Ubiquitin-like Protein ISG15|Interferon-Induced 15 kDa Protein|hUCRP|ISG15|UCRP|Interferon-Induced 17 kDa Protein|G1P2|Ubiquitin Cross-Reactive Protein
- Protein/peptide name:Interferon-stimulated Gene 15
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:18.2 kD
- Sequence:Gly2-Gly157
- Formulation:Supplied as a 0.2 um filtered solution of 50mM HEPES, 100mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-932
- Supplier No.:92-169
Specifications
About this item
Ubiquitin-Like Protein ISG15 (ISG15) is a ubiquitin-like protein that becomes conjugated to many cellular proteins upon activation by interferon-alpha and -beta. Several functions have been ascribed to the encoded protein, including chemotactic activity towards neutrophils, direction of ligated target proteins to intermediate filaments, cell-to-cell signaling, and antiviral activity during viral infections. While conjugates of this protein have been found to be noncovalently attached to intermediate filaments, this protein is sometimes secreted. ISG15 becomes conjugated to a diverse set of proteins after IFN-alpha/beta stimulation or microbial challenge. The functions or biochemical consequences ISG15 conjugation to proteins are not yet known, but it appears that this modification does not target proteins for proteasomal degradation. ISG15 shows specific chemotactic activity towards neutrophils and activates them to induce release of eosinophil chemotactic factors. Upon interferon treatment, ISG15 can be detected in both free and conjugated forms, and is secreted from monocytes and lymphocytes where it can function as a cytokine. In the cell, ISG15 co-localizes with intermediate filaments and ISGylation may modulate the JAK-STAT pathway or certain aspects of neurological disease.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.