Order Entry
ContactUsLinkComponent
Human Recombinant UBE2V1 (from E. coli)
Human Recombinant UBE2V1 (from E. coli)
Catalog #: 75790-924
Supplier:  Prosci
CAS Number:  
Human Recombinant UBE2V1 (from E. coli)
Catalog #: 75790-924
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    UEV1|UBE2V|P/OKcl.19|CROC-1|TRAF6-Regulated IKK Activator 1 Beta Uev1A|Ubiquitin-Conjugating Enzyme E2 Variant 1|UEV-1|Uev1a|CROC1|UBE2V1
  • Protein/peptide name:
    UBE2V1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Ala2-Asn147
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM HEPES, 100mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-924
  • Supplier No.:
    92-165

Specifications

About this item

Ubiquitin-Conjugating Enzyme Variant 1a (UBE2V1) is a member of the Ubiquitin-conjugating (E2) enzyme family. The E2 catalytic core domain of UBE2V1 lacks an active site cysteine residue, rendering it catalytically inactive on its own. However, in the cytoplasm UBE2V1 is able to form a catalytically active complex with UBE2N/Ubc13, which mediates the synthesis Lys63-linked Ubiquitin chains and is required for NF-kappa B activation. UBE2V1 is required for UBE2N (Ubc13)/UBE2V1 Complex-dependent Lys63-linked Ubiquitin chain formation. More specifically, UBE2V1 orients the Ubiquitin molecule to favor linkage at Lys63 via a non-covalent interaction with the Ubiquitin molecule. The UBE2V1-UBE2N heterodimer catalyzes the synthesis of non-canonical poly-ubiquitin chains that are linked through Lys63. This type of poly-ubiquitination activates IKK and does not seem to involve protein degradation by the proteasome. UBE2V1 plays a role in the activation of NF-kappa-B mediated by IL1B, TNF, TRAF6, and TRAF2. It mediates transcriptional activation of target genes. UBE2V1 also controls the progress through the cell cycle and differentiation, the error-free DNA repair pathway and contributes to the survival of cells after DNA damage.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought