Order Entry
ContactUsLinkComponent
Human Recombinant CLIC1 (from E. coli)
Human Recombinant CLIC1 (from E. coli)
Catalog #: 75790-894
Supplier:  Prosci
CAS Number:  
Human Recombinant CLIC1 (from E. coli)
Catalog #: 75790-894
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    G6|hRNCC|Nuclear Chloride Ion Channel 27|CLIC1|NCC27|Regulatory Nuclear Chloride Ion Channel Protein|Chloride Channel ABP|Chloride Intracellular Channel Protein 1
  • Protein/peptide name:
    CLIC1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29 kD
  • Sequence:
    Met1-Lys241
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-894
  • Supplier No.:
    92-150

Specifications

About this item

Chloride Intracellular Channel Protein 1 (CLIC1) belongs to the Chloride Channel CLIC family and contains one GST C-terminal domain. CLIC1 can be expressed in various cell types, but it is especially prominent in the heart, placenta, liver, kidney, and pancreas. CLIC1 can insert into membranes and form chloride ion channels. The channel activity depends on the pH. CLIC1 membrane insertion seems to be redox-regulated and may occur only under oxydizing conditions. CLIC1 is also involved in the regulation of the cell cycle.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought