Order Entry
ContactUsLinkComponent
Human Recombinant Actin-related 8 (from E. coli)
Human Recombinant Actin-related 8 (from E. coli)
Catalog #: 75790-854
Supplier:  Prosci
CAS Number:  
Human Recombinant Actin-related 8 (from E. coli)
Catalog #: 75790-854
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    ACTR8|hArp8|INO80 Complex Subunit N|Actin-Related Protein 8|ARP8|INO80N
  • Protein/peptide name:
    Actin-related 8
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.1 kD
  • Sequence:
    Met1-Trp329
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,1mM DTT,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-854
  • Supplier No.:
    92-130

Specifications

About this item

Actin-Related Protein 8 (ACTR8) is a member of the Actin family. ACTR8 is the first example in actin family that was found to be associated with mitotic chromosomes. ACTR8 plays a vital role in the functional organization of mitotic chromosomes. This protein is a proposed core component of the chromatin remodeling INO80 complex that is involved in transcriptional regulation, DNA replication and probable DNA repair, and it is required for the recruitment of INO80 (and probably the INO80 complex) to sites of DNA damage.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought