Order Entry
ContactUsLinkComponent
Human Recombinant dUTP Pyrophosphatase (from E. coli)
Human Recombinant dUTP Pyrophosphatase (from E. coli)
Catalog #: 75790-770
Supplier:  Prosci
CAS Number:  
Human Recombinant dUTP Pyrophosphatase (from E. coli)
Catalog #: 75790-770
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    dUTP Pyrophosphatase
  • Enzyme synonyms:
    Deoxyuridine 5'-Triphosphate Nucleotidohydrolase Mitochondrial, dUTPase, dUTP Pyrophosphatase, DUT
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.7 kD
  • Sequence:
    Met1-Asn164
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Used for biological assays
  • Cat. no.:
    75790-770
  • Supplier No.:
    92-088

Specifications

About this item

Deoxyuridine 5'-Triphosphate Nucleotidohydrolase Mitochondrial (dUTPase) belongs to the dUTPase family

This recombinant protein can be used for biological assays. For research use only.

dUTPase exits as a homotrimer and is involved in nucleotide metabolism. dUTPase produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA. The dUTPase increase in PCR product yield, length and fidelity enables further down-stream applications. These effects make dUTPase useful in PCR fidelity and yield-sensitive applications. dUTPase is specific for dUTP and is critical for the fidelity of DNA replication and repair.

Customers Who Bought This Also Bought