To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Signal Transducer and Activator of Transcription 3|Acute-Phase Response Factor|STAT3|APRF
- Protein/peptide name:STAT3
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:21.8 kD
- Sequence:Met1-Asn175
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-764
- Supplier No.:92-085
Specifications
About this item
Signal Transducer and Activator of Transcription 3 (STAT3) belongs to the transcription factor STAT family. STAT3 contains one SH2 domain and is a transcription factor expressed in most cell types. STAT3 is activated by multiple cytokines and growth factors including: IFN-a, IL-10, IL-6, IL-11, IL-12, IL-2, EGF etc. STAT3 functions as signal transducer and transcription activator that mediates cellular responses to interleukins, KITLG/SCF and other growth factors. In addition, STAT3 may also mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.