Order Entry
ContactUsLinkComponent
Human Recombinant Transcriptional Repressor CTCF (from E. coli)
Human Recombinant Transcriptional Repressor CTCF (from E. coli)
Catalog #: 75790-748
Supplier:  Prosci
CAS Number:  
Human Recombinant Transcriptional Repressor CTCF (from E. coli)
Catalog #: 75790-748
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    CTCF|CCCTC-Binding Factor|11-Zinc Finger Protein|CTCFL Paralog|Transcriptional Repressor CTCF
  • Protein/peptide name:
    Transcriptional Repressor CTCF
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.9 kD
  • Sequence:
    Met1-Ile154
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-748
  • Supplier No.:
    92-077

Specifications

About this item

Transcriptional Repressor CTCF (CTCF) belongs to the CTCF Zinc-Finger Protein family. CTCF contains twelve C2H2-type zinc fingers and interacts with CHD8. CTCF is widely expressed in many tissues, and it is absent in primary spermatocytes. CTCF is involved in transcriptional regulation by binding to chromatin insulators and preventing interaction between promoter and nearby enhancers and silencers. CTCF plays an essential role in oocyte and preimplantation embryo development by activating or repressing transcription. In addition, CTCF is also indispensable in the epigenetic regulation and chromatin remodeling.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought