To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:PPIase D
- Enzyme synonyms:Peptidyl-Prolyl Cis-Trans Isomerase D, PPIase D, 40 kDa Peptidyl-Prolyl Cis-Trans Isomerase, Cyclophilin-40, CYP-40, Cyclophilin-Related Protein, Rotamase D, PPID, CYP40, CYPD
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:43.9 kD
- Sequence:Met1-Ala370
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Used for biological assays
- Cat. no.:75790-704
- Supplier No.:92-055
Specifications
About this item
Peptidyl-Prolyl Cis-Trans Isomerase D (PPID) belongs to the cyclophilin-type PPIase family and PPIase D subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag
PPID is widely expressed and it contains one PPIase cyclophilin-type domain and three TPR repeats. PPID catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins. PPID can bind to the immunosuppressant cyclosporine A and is known that its overexpression suppresses the apoptosis in cancer cells.