Order Entry
ContactUsLinkComponent
Human Recombinant Thioredoxin (from E. coli)
Human Recombinant Thioredoxin (from E. coli)
Catalog #: 75790-684
Supplier:  Prosci
CAS Number:  
Human Recombinant Thioredoxin (from E. coli)
Catalog #: 75790-684
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    TRDX|Trx|Thioredoxin|ATL-Derived Factor|TXN|Surface-Associated Sulphydryl Protein|TRX1|SASP|ADF
  • Protein/peptide name:
    Thioredoxin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    13.9 kD
  • Sequence:
    Met1-Val105
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,PH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays
  • Cat. no.:
    75790-684
  • Supplier No.:
    92-045

Specifications

About this item

Thioredoxin (TXN) is a member of the Thioredoxin family. Thioredoxin exists as a disulfide-linked homodimer and contains one Thioredoxin domain. Thioredoxin is up-regulated by ionizing radiation. Thioredoxin participates in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyzes dithiol-disulfide exchange reactions. Thioredoxin also plays a role in the reversible S-nitrosylation of cysteine residues in target proteins, and thereby contributes to the response to intracellular nitric oxide.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought