Order Entry
ContactUsLinkComponent
Human Recombinant beta-Actin (from E. coli)
Human Recombinant beta-Actin (from E. coli)
Catalog #: 75790-600
Supplier:  Prosci
CAS Number:  
Human Recombinant beta-Actin (from E. coli)
Catalog #: 75790-600
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    ACTB|Actin Cytoplasmic 1|Beta-Actin
  • Protein/peptide name:
    beta-Actin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    42.8 kD
  • Sequence:
    Asp2-Phe375
  • Formulation:
    Supplied as a 0.2 um filtered solution of 10mM TrisHCl, 0.1% TritonX-100, 2mM DTT, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-600
  • Supplier No.:
    91-982

Specifications

About this item

Actins are ubiquitous globular and highly conserved proteins that are involved in various types of cell motility, structure, and integrity. Three main groups of actin isoforms, alpha, beta and gamma have been identified. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. ACTB is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to 4 others.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought