Order Entry
ContactUsLinkComponent
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from Cells)
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from Cells)
Catalog #: 75790-532
Supplier:  Prosci
CAS Number:  
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from Cells)
Catalog #: 75790-532
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Fructose-1,6-Bisphosphatase 1
  • Enzyme synonyms:
    Fructose-1, 6-bisphosphatase 1, D-fructose-1, 6-bisphosphate 1-phosphohydrolase 1, FBP, FBPase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37.8 kD
  • Sequence:
    Ala2-Gln338
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 200 mM NaCl,1 mM DTT, 10% Glycerol,pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-532
  • Supplier No.:
    91-948

Specifications

About this item

Fructose-1,6-bisphosphatase 1(FBP1) is a homotetramer protein and belongs to the FBPase class 1 family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

It involves in carbohydrate biosynthesis; gluconeogenesis pathway. FBP1 is a gluconeogenesis regulatory protein which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. FBP1 deficiency is associated with hypoglycemia and metabolic acidosis. FBP1 regulates mouse endogenous glucose production. FBP1 coupled with phosphofructokinase (PFK) takes part in the metabolism of pancreatic islet cells.

Customers Who Bought This Also Bought