Order Entry
ContactUsLinkComponent
Human Recombinant GDNF Receptor alpha 1 (from Cells)
Human Recombinant GDNF Receptor alpha 1 (from Cells)
Catalog #: 75790-488
Supplier:  Prosci
CAS Number:  
Human Recombinant GDNF Receptor alpha 1 (from Cells)
Catalog #: 75790-488
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    RET Ligand 1|RETL1|TGF-Beta-Related Neurotrophic Factor Receptor 1|GDNFRA|GFRA1|GDNF Receptor Alpha-1|GDNF Family Receptor Alpha-1|GFR-Alpha-1|GDNFR-Alpha-1|TRNR1
  • Protein/peptide name:
    GDNF Receptor alpha 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    72.3 kD
  • Sequence:
    Asp25-Lys429
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-488
  • Supplier No.:
    91-926

Specifications

About this item

Glial Cell Line-Derived Neurotrophic Factor Family Receptor alpha-1 (GDNFR alpha1) is a glycosylphosphatidylinositol (GPI) linked cell surface protein belonging to GDNF-family receptor alpha subtype which consists of at least four members. GFR alpha1and GFR alpha2 are the cognate co-receptor for the neurotrophic factor neurturin mediating the NRTN-induced autophosphorylation and activation of the RET tyrosine kinase receptor. Soluble GFR alphas released enzymatically from the cell surface by phosphatidylinositol phospholipase C, as well as recombinantly produced soluble GFR alpha1, can also bind with high affinity to GDNF and trigger the activation of Ret tyrosine kinase. Human GFR alpha1 shares 93% amino acid identity with mouse GFR alpha1.The expression of the various GFR alphas are differentially regulated in the central and peripheral nervous system, suggesting complementary roles for the GFR alphas in mediating the activities of the GDNF family of neurotrophic factors.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought