Order Entry
ContactUsLinkComponent
Human Recombinant Endoplasmic Reticulum Resident 44 (from Cells)
Human Recombinant Endoplasmic Reticulum Resident 44 (from Cells)
Catalog #: 75790-482
Supplier:  Prosci
CAS Number:  
Human Recombinant Endoplasmic Reticulum Resident 44 (from Cells)
Catalog #: 75790-482
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    KIAA0573|Thioredoxin domain-containing protein 4|ER protein 44|ER resident protein 44|ERp44TXNDC4|TXNDC4
  • Protein/peptide name:
    Endoplasmic Reticulum Resident 44
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    44.2 kD
  • Sequence:
    Glu30-Asp402
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM Tris HCl,10%glycerol,PH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-482
  • Supplier No.:
    91-923

Specifications

About this item

Endoplasmic reticulum resident protein 44 (TXNDC4) is is a 406 amino acid protein that contains one thioredoxin domain. TXNDC4 mediates thiol-dependent retention in the early secretory pathway and forms mixed disulfides with substrate proteins through its conserved CRFS motif.It can inhibit the calcium channel activity of ITPR1.It may have a role in the control of oxidative protein folding in the endoplasmic reticulum.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought