Order Entry
ContactUsLinkComponent
Human Recombinant Ephrin Type-B Receptor 1 (from Cells)
Human Recombinant Ephrin Type-B Receptor 1 (from Cells)
Catalog #: 75790-408
Supplier:  Prosci
CAS Number:  
Human Recombinant Ephrin Type-B Receptor 1 (from Cells)
Catalog #: 75790-408
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    EPHB1|Neuronally-Expressed EPH-Related Tyrosine Kinase|NET|EPH-Kike Kinase 6|EPH Tyrosine Kinase 2|Tyrosine-Protein Knase Receptor EPH-2|EK6|hEK6|EPHT2|ELK|Ephrin Type-B Receptor 1
  • Protein/peptide name:
    Ephrin Type-B Receptor 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    85.5 kD
  • Sequence:
    Met18-Pro540
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-408
  • Supplier No.:
    91-886

Specifications

About this item

Ephrin Type-B Receptor 1 (EPHB1) is a single-pass type I membrane protein that belongs to the Ephrin-B family of receptor tyrosine kinases involved in the development of embryonic nervous and vascular systems. EPHB1 contains two fibronectin type-III domains, one protein kinase domain and one Sterile Alpha Motif (SAM)domain. EPHB1 is able to stimulate fibroblast motility on extracellular matrix in a kinase-dependent manner, which is also correlated with its association with Grb7, an adaptor molecule implicated in the regulation of cell migration. It binds to Ephrin-B1, Ephrin-B2 and Ephrin-B3. EPHB1 plays an important roles in diverse biological processes including nervous system development, angiogenesis, and neural synapsis formation and maturation and may be involved in cell-cell interactions in the nervous system.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought