Order Entry
ContactUsLinkComponent
Human Recombinant IFN alpha (from Cells)
Human Recombinant IFN alpha (from Cells)
Catalog #: 75790-406
Supplier:  Prosci
CAS Number:  
Human Recombinant IFN alpha (from Cells)
Catalog #: 75790-406
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    IFN-Alpha Binding Protein|Interferon Alpha/Beta Receptor 2|IFNARB|IFN-Alpha/Beta Receptor 2|Interferon Alpha Binding Protein|IFNABR|IFNAR2|beta R2|IFN-R-2|Type I Interferon Receptor 2
  • Protein/peptide name:
    IFN alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    51.8 kD
  • Sequence:
    Ile27-Lys243
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays
  • Cat. no.:
    75790-406
  • Supplier No.:
    91-885

Specifications

About this item

Interferon alpha/ beta Receptor 2 (IFN- alpha/ beta R2) is a single-pass type I membrane protein which belongs to the type II cytokine receptor family. It complexes with IFN- alpha/ beta R1 to form the signaling receptor complex for the family of alpha and beta IFN subtypes. By alternative splicing, IFN- alpha/ beta R2 can exist as a secreted soluble protein or as a type I membrane protein. IFN- alpha/ beta R2 is the principal ligand binding subunit of the receptor. Ligand binding is stabilized by the subsequent association with IFN- alpha/ beta R1, resulting in the formation of a signaling ternary receptor complex. IFNAR2 was detected in most lymphocytes, monocytes, and granulocytes, although IFNAR2 expression was higher in the monocytes and granulocytes than in the lymphocytes. Among the lymphocyte subsets, IFNAR2 showed high expression in natural killer (NK) cells and low expression in T lymphocytes. Isoform 1 and isoform 3 of IFNAR2 are directly involved in signal transduction due to their interaction with the TYR kinase, JAK1. Isoform 1 also interacts with the transcriptional factors, STAT1 and STAT2. Both forms are potent inhibitors of type I IFN activity.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought