To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-Fc-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Osteoblast cadherin|OSF-4|OB-Cadherin|Cadherin-11|CDH11
- Protein/peptide name:Cadherin-11
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:93.5 kD
- Sequence:Phe23-Thr617
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-270
- Supplier No.:91-809
Specifications
About this item
Cadherin-11, also known as OSF-4, Osteoblast cadherin and CDH11, is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Cadherins interact with themselves in a homophilic manner in connecting cells, may thus contribute to the sorting of heterogeneous cell types. Cadherin-11 contains five cadherin domains and is mainly expressed in brain. Mature cadherin proteins consists of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. It is shown that cadherin-11 is a viable molecular target for therapeutic intervention in Glioblastoma multiforme.
- Fusion-Tag: C-Fc-6 His tag
This recombinant protein can be used for biological assays. For research use only.