To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Alkaline Phosphatase
- Enzyme synonyms:Alkaline phosphatase, placental type, also known as Alkaline phosphatase Regan isozyme, Placental alkaline phosphatase 1, ALPP and PLAP, belongs to the alkaline phosphatase family., ALPP, PLAP
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:53.7 kD
- Sequence:Ile23-Asp506
- Formulation:Supplied as a 0.2 µm filtered solution of PBS, pH 7.4 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-242
- Supplier No.:91-795
Specifications
About this item
ALPP is a membrane protein and exits as a homodimer
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
ALPP is expressed only in normal term placenta, endocervix and fallopian tube and also in ovarian and proximal gastrointestinal tumors. It has been shown to play a role in a number of processes including cell signaling, long-term potentiation, and cell adhesion, however, the best known and most commonly studied role is implicated in Alzheimer's research.