Order Entry
ContactUsLinkComponent
Podoplanin Recombinant Protein, Prosci
Podoplanin Recombinant Protein, Prosci
Catalog #: 75790-228
Supplier:  Prosci
CAS Number:  
Podoplanin Recombinant Protein, Prosci
Catalog #: 75790-228
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    PDPN|T1A|Glycoprotein 36|PA2.26 Antigen|T1-Alpha|Aggrus|Gp36|Podoplanin
  • Protein/peptide name:
    Podoplanin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    12.1 kD
  • Sequence:
    Ala23-Leu131
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-228
  • Supplier No.:
    91-788

Specifications

About this item

Podoplanin is a type-1 transmembrane protein that belongs to Podoplanin family. PDPN expressed in various specialized cell types throughout the body. It highly expressed in placenta, lung, skeletal muscle and brain, weakly expressed in brain, kidney and liver. In placenta, PDPN expressed on the apical plasma membrane of endothelium, in lung, expressed in alveolar epithelium. PDPN physiological function is related to its mucin-type character. PDPN may be involved in cell migration and/or actin cytoskeleton organization. When expressed in keratinocytes, induces changes in cell morphology with transfected cells showing an elongated shape, numerous membrane protrusions, and major reorganization of the actin cytoskeleton, increased motility and decreased cell adhesion. It requires for normal lung cell proliferation and alveolus formation at birth and Induces platelet aggregation. Nevertheless, it doesn’t have any effect on amino acid transport and the aquaporin-type water channels.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought