To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:Cell Growth Regulatory Gene 11 Protein|Cell Growth Regulator with EF Hand Domain Protein 1|Hydrophobestin|CGR11|CGREF1
- Protein/peptide name:CGREF1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:30.9 kD
- Sequence:Ala20-Ile301
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl,150mM NaCl,1mM GaCl2,pH7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-206
- Supplier No.:91-777
Specifications
About this item
Cell Growth Regulator with EF Hand Domain Protein 1 (CGREF1) is a secreted calcium ion binding protein. CGREF1 contains two EF-hand domains and both EF-hands are required for function. Human CGREF1 is synthesized as a 301 amino acid precursor that contains a 19 amino acid signal sequence, and a 282 amino acid mature chain. CGREF1 is probably digested extracellularly by an unknown serine protease generating extremely hydrophobic bioactive peptides. CGREF1 mediates cell-cell adhesion in a calcium-dependent manner. In addition, CGREF1 is able to inhibit growth in several cell lines.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.