Order Entry
ContactUsLinkComponent
Human Recombinant ER Resident 27 (from Cells)
Human Recombinant ER Resident 27 (from Cells)
Catalog #: 75790-186
Supplier:  Prosci
CAS Number:  
Human Recombinant ER Resident 27 (from Cells)
Catalog #: 75790-186
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Endoplasmic Reticulum Resident Protein 27|ERp27|ER Protein 27|C12orf46
  • Protein/peptide name:
    ER Resident 27
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    28.79 kD
  • Sequence:
    Glu26-Leu273
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-186
  • Supplier No.:
    91-767

Specifications

About this item

Endoplasmic reticulum resident protein 27, also known as ER protein 27, C12orf46 and ERP27, is an endoplasmic reticulum luminal protein which is a member of the protein disulfide isomerase family. ERP27 contains one thioredoxin domain and does not contain a CXXC active site motif. ERP27 is widely expressed in many tissues; it has highest expression in pancreas, with lower levels in spleen, lung, kidney, thymus, and bone marrow. ERP27 interacts with PDIA3 and binds somatostatin-14 via hydrophobic interactions. ERP27 may act as a protease, protein disulfide isomerase, thiol-disulfide oxidase or phospholipase.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought