Order Entry
ContactUsLinkComponent
Human Recombinant Lymphocyte Antigen 6H (from Cells)
Human Recombinant Lymphocyte Antigen 6H (from Cells)
Catalog #: 75790-182
Supplier:  Prosci
CAS Number:  
Human Recombinant Lymphocyte Antigen 6H (from Cells)
Catalog #: 75790-182
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Ly-6H|LY6H|Lymphocyte Antigen 6H
  • Protein/peptide name:
    Lymphocyte Antigen 6H
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    10.9 kD
  • Sequence:
    Leu26-Gly115
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-182
  • Supplier No.:
    91-765

Specifications

About this item

Lymphocyte Antigen 6H (LY6H) is a novel member of the LY6 family of glycosylphosphatidylinositol-anchored cell surface glycoproteins. LY6H contains one UPAR/Ly6 domain. Human LY6H is synthesized as a 140 amino acid precursor that contains a 25 amino acid signal sequence, 20 amino acid propeptide that is removed in the mature form, and a 90 amino acid mature chain. LY6H is highly expressed in the brain (cerebral cortex, amygdala, hippocampus and subthalamic nucleus) and in acute human leukemic cell line MOLT-3. It is also found in lower levels in testis, pancreas, small intestine and colon. It has been shown that LY6H may play a role in both the central nervous system and the immune system.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought