Order Entry
ContactUsLinkComponent
Human Recombinant PPIase FKBP7 (from Cells)
Human Recombinant PPIase FKBP7 (from Cells)
Catalog #: 75788-628
Supplier:  Prosci
CAS Number:  
Human Recombinant PPIase FKBP7 (from Cells)
Catalog #: 75788-628
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    PPIase FKBP7
  • Enzyme synonyms:
    Peptidyl-Prolyl Cis-Trans Isomerase FKBP7, PPIase FKBP7, 23 kDa FK506-Binding Protein, 23 kDa FKBP, FKBP-23, FK506-Binding Protein 7, FKBP-7, Rotamase, FKBP7, FKBP23
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.94 kD
  • Sequence:
    Phe23-Val423
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl,1 mM GaCl₂, 10% Glycerol,pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75788-628
  • Supplier No.:
    91-742

Specifications

About this item

Peptidyl-Prolyl Cis-Trans Isomerase FKBP7 (FKBP7) is a member of the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

FKBP7 contains two EF-hand domains and one PPIase FKBP-type domain. FKBP7 exhibits PPIase activity and function as molecular chaperones. In addition, FKBP7 accelerates the folding of proteins during protein synthesis. It has been shown that Hsp90 complex to the nucleus bind its PPIase domain to cytoplasmic dynein, the motor protein responsible for retrograde movement along microtubules.

Customers Who Bought This Also Bought