Order Entry
ContactUsLinkComponent
Human Recombinant Lysozyme G-Like 2 (from Cells)
Human Recombinant Lysozyme G-Like 2 (from Cells)
Catalog #: 75790-098
Supplier:  Prosci
CAS Number:  
Human Recombinant Lysozyme G-Like 2 (from Cells)
Catalog #: 75790-098
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    LYG2|LYGH|Lysozyme G-Like Protein 2
  • Protein/peptide name:
    Lysozyme G-Like 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    22.55 kD
  • Sequence:
    Ser20-Phe212
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,10%Glycerol,pH7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75790-098
  • Supplier No.:
    91-719

Specifications

About this item

Lysozyme G-Like Protein 2 (LYG2) is a secreted protein that belongs to the glycosyl hydrolase 23 family. LYG2 contains one SLT domain, one protein domain present in bacterial lytic transglycosylase (SLT) and in eukaryotic lysozymes (GEWL). SLT domain catalyzes the cleavage of the beta -1,4-glycosidic bond between N-acetylmuramic acid (MurNAc) and N-acetyglucosamine (GlcNAc). LYG2 has hydrolase activity which acting on glycosyl bonds, and possess lysozyme activity.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought