To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Enzyme name:Trans-2-Enoyl-CoA Reductase
- Enzyme synonyms:Trans-2-Enoyl-CoA Reductase Mitochondrial, Nuclear Receptor-Binding Factor 1, HsNrbf-1NRBF-1, MECR, NBRF1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:35.7 kD
- Sequence:Pro54-Met373
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75790-020
- Supplier No.:91-680
Specifications
About this item
Trans-2-Enoyl-CoA Reductase Mitochondrial (MECR) belongs to the zinc-containing alcohol dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
MECR localizes to the mitochondrion. It is highly expressed in skeletal and heart muscle and expressed at lower levels in the placenta, liver, kidney and pancreas, with weakly or no expression in the lung. MECR exists as a homodimer, which catalyzes the reduction of trans-2-enoyl-CoA to acyl-CoA with chain length from C6 to C16 in an NADPH-dependent manner with preference to medium chain length substrate. MECR may take part in the mitochondrial synthesis of fatty acids.