Order Entry
ContactUsLinkComponent
Mouse Recombinant IL10 (from E. coli)
Mouse Recombinant IL10 (from E. coli)
Catalog #: 75789-840
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL10 (from E. coli)
Catalog #: 75789-840
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    IL-10|Cytokine synthesis inhibitory factor|Il10|Interleukin-10|CSIF
  • Protein/peptide name:
    IL10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.9 kD
  • Sequence:
    Ser19-Ser178
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-840
  • Supplier No.:
    91-582

Specifications

About this item

Mouse Il10 is the prototypic member of the IL-10 cytokine family, including IL-10, IL-19, IL-20, IL-22 (IL-TIF), IL-24 and IL-26. Many viruses encode viral members of the IL-10 family, such as Epstein-Barr virus (EBV) and human cytomegalovirus (HCMV).Its main function is inhibiting the synthesis of a number of cytokines, including IFN-gamma, IL-2, IL-3, TNF and GM-CSF produced by activated macrophages and by helper T-cells. Although human and mouse IL-10 are 81% identical at the nucleotide and amino acid level, mouse IL-10 is species-specific and does not act on human cells. Interestingly, Human IL-10 is active on mouse cells.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought