Order Entry
ContactUsLinkComponent
TNF RII Recombinant Protein, Prosci
TNF RII Recombinant Protein, Prosci
Catalog #: 75789-834
Supplier:  Prosci
CAS Number:  
TNF RII Recombinant Protein, Prosci
Catalog #: 75789-834
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    CD120b|Tnfrsf1b|Tumor necrosis factor receptor superfamily member 1b
  • Protein/peptide name:
    TNF Receptor II
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.4 kD
  • Sequence:
    Val23-Gly258
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-834
  • Supplier No.:
    91-579

Specifications

About this item

Tumor Necrosis Factor Receptor Superfamily Member 1B (TNFRSF1B) is a member of the Tumor Necrosis Factor Receptor Superfamily. TNFRSF1B contains four TNFR-Cys repeats. TNFRSF1B can be cleaved into the following 2 chains: Tumor necrosis factor receptor superfamily member 1b and membrane form and Tumor necrosis factor-binding protein 2. TNFRSF1B is a receptor with high affinity for TNFSF2/TNF- alpha and approximately 5-fold lower affinity for homotrimeric TNFSF1/lymphotoxin- alpha. TNFRSF1B mediates most of the metabolic effects of TNF- alpha. TNF- alpha-induced apoptosis suggests that it regulates TNF- alpha function by antagonizing its biological activity.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought