Order Entry
ContactUsLinkComponent
Human Recombinant SPESP1 (from Cells)
Human Recombinant SPESP1 (from Cells)
Catalog #: 75789-816
Supplier:  Prosci
CAS Number:  
Human Recombinant SPESP1 (from Cells)
Catalog #: 75789-816
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Sperm Equatorial Segment Protein 1|SPESP1|Glycosylated 38 kDa Sperm Protein C-7/8|ESP|SP-ESP|Equatorial Segment Protein
  • Protein/peptide name:
    SPESP1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37.9 kD
  • Sequence:
    Tyr20-Tyr350
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-816
  • Supplier No.:
    91-570

Specifications

About this item

Sperm Equatorial Segment Protein 1 (SPESP1) is a member of the SPESP1 family. SPESP1 is highly expressed in the testis, where it is localized to the acrosome of postmeiotic stages of spermiogenesis; it is expressed at lower levels in the placenta and fetal lung. SPESP1 is involved in the multicellular organisimal development. Disruption of SPESP1 leads to abnormal distribution of sperm proteins resulting in a detached membrane from the equatorial segment and less fertile sperm. SPESP1 may interact with IZUMO1 and MN9 antigen and it contains an N-glycosylation site as well as several cAMP-dependent kinase, protein kinase C, and casein kinase II consensus phosphorylation sites.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought