Order Entry
ContactUsLinkComponent
Human Recombinant ST6GalNAc2 (from Cells)
Human Recombinant ST6GalNAc2 (from Cells)
Catalog #: 75789-800
Supplier:  Prosci
CAS Number:  
Human Recombinant ST6GalNAc2 (from Cells)
Catalog #: 75789-800
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    GalNAc Alpha-2|ST6GALNAC2|SThM|ST6GalNAc II|ST6GalNAcII|6-Sialyltransferase II|Sialyltransferase 7B|Alpha-N-Acetylgalactosaminide Alpha-2|SIAT7-B|6-Sialyltransferase 2|SIAT7B|SIATL1
  • Protein/peptide name:
    ST6GalNAc2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    38.84 kD
  • Sequence:
    Ser29-Arg374
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-800
  • Supplier No.:
    91-562

Specifications

About this item

Alpha-N-Acetylgalactosaminide alpha-2,6-Sialyltransferase 2 (ST6GALNAC2) belongs to the glycosyltransferase 29 family that adds sialic acids to the non-reducing ends of glycoconjugates. At the cell surface, these modifications play roles in cell-cell and cell-substrate interactions, bacterial adhesion and protein targeting. ST6GALNAC2 is localized to the Golgi apparatus membrane. ST6GALNAC2 is highly expressed in lactating mammary glands and the adult testis; it is expressed at lower levels in the kidney. ST6GALNAC2 can catalyze 2,6-sialylation of the Tn antigen.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought