Order Entry
ContactUsLinkComponent
Human Recombinant CLEC10A (from Cells)
Human Recombinant CLEC10A (from Cells)
Catalog #: 75789-724
Supplier:  Prosci
CAS Number:  
Human Recombinant CLEC10A (from Cells)
Catalog #: 75789-724
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Macrophage Lectin 2|C-Type Lectin Domain Family 10 Member A|C-Type Lectin Superfamily Member 14|CD301|CLEC10A|HML|CLECSF14|CLECSF13
  • Protein/peptide name:
    CLEC10A
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    29.8 kD
  • Sequence:
    Gln61-His316
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-724
  • Supplier No.:
    91-524

Specifications

About this item

C-Type Lectin Domain Family 10 Member A (CLEC10A) is a type II transmembrane C-type lectin that is expressed on immature myleloid dendritic cells and alternatively activated (tolerogenic) macrophages. CLEC10A/MGL binds and internalizes molecules with terminal nonsialylated GalNAc carbohydrates such as the Tn carcinoma antigen. CLEC10A/MGL also binds the GP envelope glycoprotein on Marburg and Ebola viruses and enhances viral entry and infectivity. It constitute a unique class of C-type lectins because of their specificity for galactose and its structural homologues. CLEC10A is thought to participate in the recognition of molecules from both altered self and pathogens due to its monosaccharide specificity for Gal and N-acetylgalactosamine (GalNAc). Human and rat carry a single gene for CLEC10A/MGL, while mouse has two closely related MGL1 and MGL2 genes.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought