Order Entry
ContactUsLinkComponent
Human Recombinant Protein Disulfide-Isomerase A6 (from Cells)
Human Recombinant Protein Disulfide-Isomerase A6 (from Cells)
Catalog #: 75789-628
Supplier:  Prosci
CAS Number:  
Human Recombinant Protein Disulfide-Isomerase A6 (from Cells)
Catalog #: 75789-628
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    Protein Disulfide-Isomerase A6
  • Enzyme synonyms:
    Protein Disulfide-Isomerase A6, Endoplasmic Reticulum Protein 5, ER Protein 5, ERp5, Protein Disulfide Isomerase P5, Thioredoxin Domain-Containing Protein 7, PDIA6, P5, TXNDC7
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.2 kD
  • Sequence:
    Leu20-Leu440
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-628
  • Supplier No.:
    91-476

Specifications

About this item

Protein Disulfide-Isomerase A6 (PDIA6) is a 48.5kDa protein that belongs to the protein disulfide isomerase family (PDI)

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

PDIA6 is an enzyme in the endoplasmic reticulum in eukaryotes which catalyzes the formation and breakage of disulfide bonds between cysteine residues within proteins as they fold. The PDIA6 expressed in platelets, its functions as a chaperone that inhibits aggregation of misfolded proteins. PDIA6 is part a large chaperone multiprotein complex comprising DNAJB11, HSP90B1, HSPA5, HYOU, PDIA2, PDIA4, PDIA6, PPIB, SDF2L1, UGT1A1. PDIA6 also plays a role in platelet aggregation and activation by agonists such as convulxin, collagen and thrombin.

Customers Who Bought This Also Bought