Order Entry
ContactUsLinkComponent
Human Recombinant IL-1RAcP (from Cells)
Human Recombinant IL-1RAcP (from Cells)
Catalog #: 75789-540
Supplier:  Prosci
CAS Number:  
Human Recombinant IL-1RAcP (from Cells)
Catalog #: 75789-540
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    IL-1 Receptor Accessory Protein|IL1RAP|Interleukin-1 Receptor Accessory Protein|IL-1R3|IL-1R-3|IL1R3|IL-1RAcP|Interleukin-1 Receptor 3|C3orf13
  • Protein/peptide name:
    IL-1RAcP
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    39.75 kD
  • Sequence:
    Ser21-Gln356
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-540
  • Supplier No.:
    91-432

Specifications

About this item

Interleukin-1 Receptor Accessory Protein (IL-1RAcP) is a member of the interleukin-1 receptor family. It contains three Ig-like C2-type domains in the extracellular region and a long cytoplasmic domain implicated in signal transduction. IL-1RAcP acts as a non-ligand binding accessory component of the receptors for IL1 alpha, IL1 beta and IL33. IL-1RAcP mediates interleukin-1-dependent activation of NF-kappa-B. It is part of the membrane-bound form of the IL-1 receptor. IL-1 RAcP takes part in the Signaling ways by the formation of a ternary complex containing IL1R1, TOLLIP, MYD88, and IRAK1 or IRAK2. In addition, IL-1RAcP modulates the response to interleukins by associating with soluble IL1R1 and enhancing interleukin-binding to the decoy receptor.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought