Order Entry
ContactUsLinkComponent
Human Recombinant beta-Galactosidase (from Cells)
Human Recombinant beta-Galactosidase (from Cells)
Catalog #: 75789-532
Supplier:  Prosci
CAS Number:  
Human Recombinant beta-Galactosidase (from Cells)
Catalog #: 75789-532
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    beta-Galactosidase
  • Enzyme synonyms:
    Beta-Galactosidase, Acid Beta-Galactosidase, Lactase, Elastin Receptor 1, GLB1, ELNR1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    74.63 kD
  • Sequence:
    Leu24-Val677
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays
  • Cat. no.:
    75789-532
  • Supplier No.:
    91-428

Specifications

About this item

beta Galactosidase is a lysosomal beta Galactosidase that hydrolyzes the terminal beta Galactose from Ganglioside and Keratan sulfate

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

In lysosome, the mature beta Galactosidase protein associates with Cathepsin A and Neuraminidase 1 to form the lysosomal multienzyme complex . An alternative splicing at the RNA level of beta Galactosidase results a catalytically inactive beta Galactosidase that plays an important role in vascular development. Defects of beta -galactosidase (GLB1) are the cause of diseases like GM1-gangliosidosis which is a lysosomal storage disease and Morquio Syndrome B that cause patients to have abnormal elastic fibers. More than 100 mutations have been identified for beta Galactosidase, which result in different residual activities of the mutant enzymes and a spectrum of symptoms in the two related diseases.

Customers Who Bought This Also Bought