Order Entry
ContactUsLinkComponent
Pref-1 Recombinant Protein, Prosci
Pref-1 Recombinant Protein, Prosci
Catalog #: 75789-514
Supplier:  Prosci
CAS Number:  
Pref-1 Recombinant Protein, Prosci
Catalog #: 75789-514
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    FA1|DLK|Protein Delta Homolog 1|DLK-1|pG2|DLK1
  • Protein/peptide name:
    PREF-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    30.15 kD
  • Sequence:
    Ala24-Pro297
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-514
  • Supplier No.:
    91-419

Specifications

About this item

Protein Delta Homolog 1 (DLK-1) is a transmembrane protein which contains a signal peptide, an extracellular domain with six tandem epidermai growth factor (EGF)-like domains, a single pass transmembrane domain, and a short cytoplasmic tail. It is found within the stromal cells in close contact to the vascular structure of placental villi, yolk sac, fetal liver, adrenal cortex and pancreas and in the beta cells of the islets of Langerhans in the adult pancreas. In addition, it is detected in some forms of neuroendocrine lung tumor tissue. DLK-1 may have a improtant role in neuroendocrine differentiation.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought