To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:PLAUR|Urokinase Plasminogen Activator Surface Receptor|uPAR|MO3|Monocyte activation antigen Mo3|CD87|U-PAR
- Protein/peptide name:CD87
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:32.6 kD
- Sequence:Leu23-Arg303
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-376
- Supplier No.:91-350
Specifications
About this item
The Urokinase Type Plasminogen Activator (uPA) receptor (uPAR) is a widely expressed receptor for urokinase plasminogen activator (uPA) and pro-uPA. uPAR / CD87 is a highly glycosylated, 55-60kDa integral membrane protein linked to the plasma membrane by a glycosylphosphatidylinositol (GPI) anchor. uPAR is expressed by T-cells, NK cells, monocytes, and neutrophils as well as non-hematopoietic cells that include vascular endothelial cells, fibroblasts, smooth muscle cells, keratinocytes, placental trophoblasts, hepatocytes, and a wide variety of tumor cells (including breast, colon, and prostate carcinoma, melanoma). It plays a critical role in the regulation of cell-surface plasminogen activation in physiological and pathological conditions, and it is also involved in cellular adhesion, the transmission of extracellular signals across the plasma membrane and the subsequent regulation of gene expression. uPAR has been implicated in several biological processes including angiogenesis, monocyte migration, cancer metastasis, trophoblast implantation, and wound healing. Human uPAR is encoded as a 313 amino acid residue polypeptide, excluding a 22 residue signal peptide and shows 60-70% similarity with the murine uPAR amino acid sequence although binding of uPA to uPAR shows strong species specificity.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.