Order Entry
ContactUsLinkComponent
Human Recombinant PDGF R beta (from Cells)
Human Recombinant PDGF R beta (from Cells)
Catalog #: 75789-372
Supplier:  Prosci
CAS Number:  
Human Recombinant PDGF R beta (from Cells)
Catalog #: 75789-372
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    PDGFRB|Platelet-Derived Growth Factor Receptor 1|PDGFR|CD140 Antigen-like Family Member B|CD140b|Beta-Type Platelet-Derived Growth Factor Receptor|PDGFR1|PDGFR-Beta|Platelet-Derived Growth Factor Receptor Beta|Beta Platelet-Derived Growth Factor Receptor|PDGFR-1|PDGF-R-Beta
  • Protein/peptide name:
    PDGF R beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    57.17 kD
  • Sequence:
    Leu33-Phe530
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-372
  • Supplier No.:
    91-348

Specifications

About this item

Platelet-Derived Growth Factor Receptor beta (PDGFR- beta ) is a member of the protein kinase superfamily and CSF-1/PDGF receptor subfamily. The PDGF family consists of PDGF-A, -B, -C and -D, which form either homo- or heterodimers (PDGF-AA, -AB, -BB, -CC, -DD). The four PDGFs are inactive in their monomeric forms. The PDGFs bind to the protein tyrosine kinase receptors PDGF receptor- alpha and - beta . These two receptor isoforms dimerize upon binding the PDGF dimer, leading to three possible receptor combinations, namely - alpha alpha, - beta beta and - alpha beta . The extracellular region of the PDGF receptor- beta consists of five immunoglobulin-like domains while the intracellular part is a tyrosine kinase domain. In addition to being a potent mitogen for cells of mesenchymal origin, PDGF has also been shown to be a potent chemoattractant for mesenchymal cells, mononuclear cells, and neutrophils and has been reported to be important in the modification of cellular matrix constituents.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought