Order Entry
ContactUsLinkComponent
Human Recombinant High Mobility Group B1 (from Cells)
Human Recombinant High Mobility Group B1 (from Cells)
Catalog #: 75789-332
Supplier:  Prosci
CAS Number:  
Human Recombinant High Mobility Group B1 (from Cells)
Catalog #: 75789-332
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    HMGB1|High Mobility Group Protein B1|HMG1|High Mobility Group Protein 1|HMG-1
  • Protein/peptide name:
    High Mobility Group B1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.93 kD
  • Sequence:
    Gly2-Glu215
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, 1mM DTT, 10% Glycerol, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-332
  • Supplier No.:
    91-328

Specifications

About this item

High mobility group protein B1 is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3.It Contains 2 HMG box DNA-binding domains entitled box A and box B and It is a highly negative-charged C terminus. As a nuclear protein, HMGB1 stabilizes nucleosomes and allows bending of DNA that facilitates gene transcription which is essential for individual survival. Meanwhile, it is revealed that HMGB1 can also act as a cytokine extracellularlly and regulates monocyte, T cell, dendritic cell activities in inflammatory responses.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought