To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:ICAM2|ICAM-2|CD102|Intercellular Adhesion Molecule 2
- Protein/peptide name:ICAM2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:23.13 kD
- Sequence:Lys25-Gln223
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 2mM CaCl2, 2mM MgCl2, 5% Threhalose, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75789-248
- Supplier No.:91-286
Specifications
About this item
ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. Interactions of ICAM-2 and the integrin receptors mediate cell adhesion in a wide range of lymphocyte, monocyte, natural killer cell, and granulocytewith other cells, and play important roles in many adhesion-dependent immune and inflammation responses, such as T cell aggregation, NK-cell cytotoxicity and migration, lymphocyte recirculation, etc. Serum levels of ICAM-2 correlated significantly with the inflammatory and course sequences of trichinosis in mice and had a similar relation with blood eosinophilia. So, estimation of ICAM-2 serum levels may prove useful in diagnosis of trichinosis recent infections, and in monitoring the prognosis and response to treatment.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.