Order Entry
ContactUsLinkComponent
Human Recombinant Galectin 1 (from E. coli)
Human Recombinant Galectin 1 (from E. coli)
Catalog #: 75789-206
Supplier:  Prosci
CAS Number:  
Human Recombinant Galectin 1 (from E. coli)
Catalog #: 75789-206
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological activity:
    Measured by its ability to agglutinate human red blood cells.
  • Protein synonyms:
    Galectin-1|HBL|LGALS1|S-Lac Lectin 1|Galaptin|Beta-Galactoside-Binding Lectin L-14-I|14 kDa Lectin|Putative MAPK-Activating Protein PM12|Lectin Galactoside-Binding Soluble 1|HLBP14|HPL|Lactose-Binding Lectin 1|14 kDa Laminin-Binding Protein|Gal-1
  • Protein/peptide name:
    Galectin 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    15.78 kD
  • Sequence:
    Ala2-Asp135
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 10mM PB, 200mM NaCl, 2mM DTT, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-206
  • Supplier No.:
    91-265

Specifications

About this item

Galectin-1 is a member of growing family of evolutionary conserved animal lectins. Galectin-1 is widely expressed in many cells and tissues. Galectins consists of a Galectin domain and two Beta-galactoside binding domains. Galectin-1 can binds LGALS3BP and interacts with CD2, CD3, CD4, CD7, CD43 and CD45. Galectin-1 may act as an autocrine negative growth factor which regulates apoptosis, cell proliferation and cell differentiation. In addition, Galectin-1 plays improtant roles in immunosuppressive and antiinflammatory properties.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought