Order Entry
ContactUsLinkComponent
Human Recombinant PPIase H (from E. coli)
Human Recombinant PPIase H (from E. coli)
Catalog #: 75789-182
Supplier:  Prosci
CAS Number:  
Human Recombinant PPIase H (from E. coli)
Catalog #: 75789-182
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Enzyme name:
    PPIase H
  • Enzyme synonyms:
    Peptidyl-Prolyl Cis-Trans Isomerase H, PPIase H, Rotamase H, Small Nuclear Ribonucleoprotein Particle-Specific Cyclophilin H, CypH, U-snRNP-Associated Cyclophilin SnuCyp-20, USA-CYP, PPIH, CYP20
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.4 kD
  • Sequence:
    Met1-Met177
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-182
  • Supplier No.:
    91-233

Specifications

About this item

Peptidyl-Prolyl Cis-Trans Isomerase H (PPIH) belongs to the Cyclophilin-type PPIase family that accelerate the folding of proteins

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag

PPIases can catalyze the cis-trans isomerization of Proline Imidic peptide bonds in oligopeptides. PPIH participates in pre-mRNA splicing. It is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. In addition, PPIH has PPIase activity and may play a role as a chaperone mediating the interactions between different proteins inside the spliceosome.

Customers Who Bought This Also Bought