Order Entry
ContactUsLinkComponent
Human Recombinant Annexin A13 (from E. coli)
Human Recombinant Annexin A13 (from E. coli)
Catalog #: 75789-084
Supplier:  Prosci
CAS Number:  
Human Recombinant Annexin A13 (from E. coli)
Catalog #: 75789-084
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Annexin A13|Annexin-13|ANXA13|Intestine-Specific Annexin|ISA|ANX13|Annexin XIII
  • Protein/peptide name:
    Annexin A13
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    35.4 kD
  • Sequence:
    Gly2-His316
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75789-084
  • Supplier No.:
    91-184

Specifications

About this item

Annexin A13 (ANXA13) belongs to the annexin family which plays a role in phospholipase inhibition, cytoskeletal interactions, intracellular signal transduction pathways and regulation of cellular growth. ANXA13 contains four annexin repeats and a pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA13 is highly expressed in intestinal and kidney epithelial cells. The specific function of ANXA13 has not yet been determined; however it is associated with the plasma membrane of undifferentiated, proliferating crypt epithelial cells as well as differentiated villus enterocytes.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought