Order Entry
ContactUsLinkComponent
Human Recombinant GOT1 (from E. coli)
Human Recombinant GOT1 (from E. coli)
Catalog #: 75788-970
Supplier:  Prosci
CAS Number:  
Human Recombinant GOT1 (from E. coli)
Catalog #: 75788-970
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    Aspartate Aminotransferase Cytoplasmic|Transaminase A|GOT1|Glutamate Oxaloacetate Transaminase 1
  • Protein/peptide name:
    GOT1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    47.31 kD
  • Sequence:
    Ala2-Leu414
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 2mM DTT, 20% Glycerol, pH 7.5 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75788-970
  • Supplier No.:
    91-127

Specifications

About this item

Glutamate Oxaloacetate Transaminase 1 (GOT1) is a cytoplasmic protein. GOT1 belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family. GOT1 is a pyridoxal phosphate-dependent enzyme that exists in cytoplasmic and mitochondrial forms. GOT1 plays a key role in amino acid metabolism and the urea and tricarboxylic acid cycles. GOT1 involves in L-methionine salvage from methylthioadenosine, aspartate catabolic process, cellular response to insulin stimulus, polyamine metabolic process, and glucocorticoid stimulus.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought