Order Entry
ContactUsLinkComponent
Human Recombinant CCL26 (from E. coli)
Human Recombinant CCL26 (from E. coli)
Catalog #: 75788-918
Supplier:  Prosci
CAS Number:  
Human Recombinant CCL26 (from E. coli)
Catalog #: 75788-918
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    MIP-4-Alpha|Macrophage Inflammatory Protein 4-Alpha|TSC-1|Small-Inducible Cytokine A26|Thymic Stroma Chemokine-1|C-C Motif Chemokine 26|Eotaxin-3|SCYA26|CC Chemokine IMAC|CCL26
  • Protein/peptide name:
    CCL26
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    8.21 kD
  • Sequence:
    Ser27-Leu94
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75788-918
  • Supplier No.:
    91-097

Specifications

About this item

Chemokine (C C Motif) Ligand 26 (CCL26) is a novel small cytokine belonging to the CC chemokine family, which involved in immunoregulatory and inflammatory processes. CCL26 is expressed constitutively in thymus, but only transiently in phytohemagglutinin-stimulated peripheral blood mononuclear cells. It specifically binds and induces chemotaxis in T cells and elicits its effects by interacting with the chemokine receptor CCR4. Eotaxin-3/CCL26, along with Eotaxin-1 and Eotaxin-2, selectively activates the CC chemokine receptor 3 (CCR3). The Eotaxin-3-CCR3 interaction may play an important role in allergic diseases such as atopic dermatitis and bronchial asthma. The full-length cDNA for Eotaxin-3 encodes a protein of 94 amino acids with a putative signal peptide of either 23 or 26 amino acid residues. Both the 71 and 68 amino acid residue variants of recombinant Eotaxin-3 demonstrate equal potency in inducing chemotaxis of a human CCR3-transfected cell line. Unlike most other CC chemokines, Eotaxin-3 maps to human chromosome 7q11.2, within 40 kilobases of the Eotaxin-2 loci. Eotaxin-3 and Eotaxin-2 are unique in that they are the only chemokines identified to date that map to chromosome 7.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought