Order Entry
ContactUsLinkComponent
Human Recombinant CCL23 (from E. coli)
Human Recombinant CCL23 (from E. coli)
Catalog #: 75788-872
Supplier:  Prosci
CAS Number:  
Human Recombinant CCL23 (from E. coli)
Catalog #: 75788-872
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein synonyms:
    MPIF1|MIP3|CK-Beta-8|SCYA23|Myeloid Progenitor Inhibitory Factor 1|C-C Motif Chemokine 23|Small-Inducible Cytokine A23|MPIF-1|CKB-8|Macrophage Inflammatory Protein 3|MIP-3|CCL23
  • Protein/peptide name:
    CCL23
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    11.5 kD
  • Sequence:
    Arg22-Asn120
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 250mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays
  • Cat. no.:
    75788-872
  • Supplier No.:
    91-074

Specifications

About this item

Human Chemokine (C-C Motif) Ligand 23 (CCL23) is a small cytokine belonging to the CC chemokine family. CCL23 is also known as myeloid progenitor inhibitory factor MPIF-1, CK8 and SCYA23. CCL23 cDNA encodes a 120 amino acid residue precursor protein with a putative 21 amino acid residue signal peptide that is cleaved to generate a 99 amino acid residue mature CCL23 (amino acids 22 -120). Additional N-terminal Processing of the 99 amino acid residue variant can generate a 75 amino acid residue peptide (amino acid 46-120) that is significantly more active than the 99 amino acid residue variant. CCL23 binds to CCR1 with high affinity and has chemotactic activity for monocytes, dendritic cells, and osteoclast precursors. CCL23 enhances angiogenesis of endothelial cells, but reduces the proliferation of progenitor cells giving rise to granulocyte and monocyte lineages.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought