To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.05 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein synonyms:ILC|CC Chemokine ILC|CTACK|SCYA27|Cutaneous T-Cell-Attracting Chemokine|IL-11 R-Alpha-Locus Chemokine|Small-Inducible Cytokine A27|Eskine|C-C Motif Chemokine 27|Skinkine|CCL27
- Protein/peptide name:CCL27
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:10.1 kD
- Sequence:Phe25-Gly112
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 500mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
- Cat. no.:75788-828
- Supplier No.:91-052
Specifications
About this item
Human Chemokine (C-C Motif) Ligand 27 (CCL27) is a small cytokine that is a member of the CC chemokine family; it is expressed in numerous tissues, including gonads, thymus, placenta and skin. CCL27 elicits its chemotactic effects by binding to the chemokine receptor CCR10. Predominantly expressed in the skin, CCL27 is associated with T cell-mediated inflammation of the skin. Human and Mouse CCL27 share 84% sequence identity in the mature form.
This recombinant protein can be used for biological assays. For research use only.