Order Entry
ContactUsLinkComponent
Mouse Recombinant IL1 beta (from E. coli)
Mouse Recombinant IL1 beta (from E. coli)
Catalog #: 75788-788
Supplier:  Prosci
CAS Number:  
Mouse Recombinant IL1 beta (from E. coli)
Catalog #: 75788-788
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological activity:
    ED50 is less than 0.01 ng/ml as determined by the dose-dependent stimulation of mouse D10S cells.<br />Specific Activity of 1.0 x 10<sup>8</sup> IU/mg.
  • Protein synonyms:
    IL-1 Beta|Il1b|IL-1b|Interleukin-1 Beta|IL-1F2
  • Protein/peptide name:
    IL1 beta
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.4 kD
  • Sequence:
    Val118-Ser269
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 50mM TrisHCl, 50mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75788-788
  • Supplier No.:
    91-032

Specifications

About this item

Interleukin-1 (IL-1) designates two proteins, IL-1 alpha and IL-1 beta , which are the products of distinct genes, but recognize the same cell surface receptors. IL-1 alpha and IL-1 beta are structurally related polypeptides that show approximately 25% homology at the amino acid level. Both proteins are produced by a wide variety of cells in response to stimuli such as those produced by inflammatory agents, infections, or microbial endotoxins. The proteins are synthesized as 31 kDa precursors that are subsequently cleaved into proteins with molecular weights of approximately 17.5 kDa. The specific protease responsible for the processing of IL-1 beta , designated interleukin 1 beta -converting enzyme (ICE), has been described. Mature human and mouse IL-1 beta share approximately 75% amino acid sequence identity and human IL-1 beta has been found to be active on murine cell lines.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought