Order Entry
ContactUsLinkComponent
Human Recombinant IFN alpha 2a (from E. coli)
Human Recombinant IFN alpha 2a (from E. coli)
Catalog #: 75788-760
Supplier:  Prosci
CAS Number:  
Human Recombinant IFN alpha 2a (from E. coli)
Catalog #: 75788-760
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological activity:
    Specific Activity is greater than 1.0 x 108 IU/ mg as determined in a viral resistance assay using VSV-WISH cells.
  • Protein synonyms:
    LeIF A|IFN-Alpha-2|Interferon Alpha-A|IFNA2|Interferon Alpha-2
  • Protein/peptide name:
    IFN alpha 2a
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.24 kD
  • Sequence:
    Cys24-Glu188
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay
  • Cat. no.:
    75788-760
  • Supplier No.:
    91-018

Specifications

About this item

At least 23 different variants of IFN- alpha are known. The individual proteins have molecular masses between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. All IFN- alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN- alpha subtypes only differ in their sequences by one or two positions. Naturally occurring variants also include proteins truncated by 10 amino acids at the carboxy-terminal end.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought