Order Entry
ContactUsLinkComponent
Human Recombinant IL11 (from P. pichia)
Human Recombinant IL11 (from P. pichia)
Catalog #: 75791-998
Supplier:  Prosci
CAS Number:  
Human Recombinant IL11 (from P. pichia)
Catalog #: 75791-998
Supplier:  Prosci
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    P. pichia
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Biological activity:
    ED50 is less than 0.2 ng/ml as determined by the dose-dependent stimulation of murine 7TD1 proliferation.<br />Specific Activity of 8.0 x 10<sup>6</sup> IU/ mg.
  • Protein synonyms:
    Adipogenesis Inhibitory Factor|Interleukin-11|Oprelvekin|IL11|IL-11|AGIF
  • Protein/peptide name:
    IL11
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19 kD
  • Sequence:
    Gly23-Leu199
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 2% Glycine, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Cat. no.:
    75791-998
  • Supplier No.:
    91-005

Specifications

About this item

Interleukin 11 (IL-11) is a thrombopoietic growth factor that directly stimulates the proliferation of hematopoietic stem cells and megakaryocyte progenitor cells and induces megakaryocyte maturation resulting in increased platelet production. IL-11 is a member of a family of human growth factors that includes human growth hormone, granulocyte colony-stimulating factor, and other growth factors.

This recombinant protein can be used for biological assays. For research use only.

Customers Who Bought This Also Bought