To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:vasodilator-stimulated phosphoprotein
- Clonality:Polyclonal
- Gene ID:P50552
- Host:Rabbit
- Reactivity:Pig,Human,Mouse
- Antigen symbol:VASP
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:25 ug
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:VASP
- Storage buffer:Liquid. In PBS containing 1mg/ml BSA and 0.02% sodium azide.
- Storage temperature:Stable for at least 1 year after receipt when stored at –20 °C.
- Concentration:250 ug/mL
- Shipping temperature:4 °C
- Immunogen:Recombinant human VASP fused to a His-tag.
- Tested applications:Immunocytochemistry
- Purification:Antigen affinity purified.
- Cat. no.:75793-488
- Supplier No.:36-346
Specifications
About this item
VASP (vasodilator-stimulated phosphoprotein) is a proline-rich protein substrate of cAMP- and cGMP-dependent protein kinases. VASP is an actin-associated protein involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance, lamellipodial and filopodial dynamics, platelet activation and cell migration. VASP promotes actin filament elongation. It protects the barbed end of growing actin filaments against capping and increases the rate of actin polymerization in the presence of capping proteins. VASP stimulates actin filament elongation by promoting the transfer of profilin-bound actin monomers onto the barbed end of growing actin filaments and it plays a role in actin-based mobility of Listeria monocytogenes in host cells. It regulates actin dynamics in platelets and plays an important role in regulating platelet aggregation. VASP phosphorylation is used to monitor the effect of so-called antiplatelet drugs that reduce platelet reactivity and are used to prevent stent thrombosis, strokes and heart attacks in patients. Phosphorylation of VASP at Ser157 causes a mobility shift in SDS gel electrophoresis from 46 to 50kDa, which has been used as a convenient marker to monitor cyclic nucleotide-dependent protein kinase activity.
Immunocytochemistry: (formaldehyde fixed cells; 1:250). Immunoprecipitation: (1:125). Western Blot: 1:2500. Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Pig