To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:indoleamine 2,3-dioxygenase 1
- Clonality:Polyclonal
- Gene ID:P28776
- Host:Rabbit
- Reactivity:Mouse
- Antigen symbol:Ido1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 ug
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Ido1
- Storage buffer:Liquid. 0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage temperature:Stable for at least 6 months after receipt when stored at –20 °C.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Recombinant mouse IDO.
- Tested applications:Immunocytochemistry
- Cat. no.:75793-322
- Supplier No.:36-231
Specifications
About this item
IDO catalyzes the first and rate-limiting step in the main pathway of human tryptophan catabolism, the kynurenine pathway. Proinflammatory mediators, such as endotoxin and IFN-gamma induce the expression of IDO in several tissues. IDO-dependent suppression of T-cell responses might function as natural immunoregulatory mechanism. Physiological IDO activity has been implicated in T-cell tolerance to tumors, dysfunctional selftolerance in non-obese diabetic (NOD) mice, and as a protective negative regulator in autoimmune disorders.
ELISA: (direct and indirect: 1:2,000-1:10,000). Immunocytochemistry: (1:500-1:2,000). Immunohistochemistry: (paraffin sections (1:200-1:500)). Western Blot: (1:2,000-1:5,000 using ECL; suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk; suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: Ido1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse