To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:nicotinamide phosphoribosyltransferase
- Clonality:Polyclonal
- Gene ID:P43490
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:NAMPT
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 ug
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:NAMPT
- Storage buffer:Liquid. 0.2um-filtered solution in PBS, pH 7.4. Contains no preservatives.
- Storage temperature:Stable for at least 6 months after receipt when stored at –20 °C.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:Recombinant human Nampt (visfatin/PBEF).
- Cat. no.:75793-306
- Supplier No.:36-223
Specifications
About this item
Nampt also known as pre-B cell colony-enhancing factor 1 (PBEF1) or visfatin is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity.
ELISA: (direct and indirect: 1:2,000-1:5,000). Immunohistochemistry: (paraffin sections (1:500-1:1,000)). Western Blot: (1:2,000-1:5,000 using ECL; suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk; suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: NAMPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human